missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PIR-A/B Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5308-SP
This item is not returnable.
View return policy
Description
PIR-A/B Monoclonal specifically detects PIR-A/B in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| PIR-A/B | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| A2NTJ8 | |
| Mouse myeloma cell line NS0-derived recombinant mouse PIR-A1 Gly24-Met650 Accession # A2NTJ8 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 404127 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects mouse PIR-A and mouse PIR-B in direct ELISAs. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction