Learn More
Mouse Plexin B2 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems FAB6836G-100UG
Product Code. 18762489
  • € 518.00 / 100µg
Description
Plexin B2 Monoclonal specifically detects Plexin B2 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plexin B2
Monoclonal
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
MM1, MM1KIAA0315dJ402G11.3, Nbla00445, PLEXB2, plexin B2, plexin-B2, PLXNB2
Rat
Protein A or G purified from hybridoma culture supernatant
Primary
Detects mouse Plexin B2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Plexin B2 is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG2a
Flow Cytometry
772417
Flow Cytometry 0.25-1 μg/106 cells
B2RXS4
PLXNB2
Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin B2, Leu20-Trp1029, Accession # B2RXS4
100 μg
23654
Mouse
Supernatant