missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PLZF Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB8395
This item is not returnable.
View return policy
Description
PLZF Monoclonal specifically detects PLZF in Mouse samples. It is validated for Western Blot.
Specifications
| PLZF | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_001028496 | |
| ZBTB16 | |
| E. coli-derived recombinant mouse PLZF Met1-Gln254 Accession # NP_001028496 | |
| 100 μg | |
| Primary | |
| Detects mouse PLZF in direct ELISA and Western Blots. | |
| Mouse | |
| Purified |
| Western Blot | |
| 816416 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PLZFzinc finger and BTB domain-containing protein 16, ZBTB16, zinc finger and BTB domain containing 16, Zinc finger protein 145, zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia), Zinc finger protein PLZF, ZNF145, ZNF145Promyelocytic leukemia zinc finger protein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7704 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction