missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Pro EGF Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB5256
This item is not returnable.
View return policy
Description
EGF Monoclonal specifically detects EGF in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| EGF | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P01132 | |
| EGF | |
| Mouse myeloma cell line NS0-derived recombinant mouse EGF Trp29-Arg1029 Accession # P01132 | |
| 100 μg | |
| Primary | |
| Detects mouse Pro EGF in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Amphiregulin, rmEpigen, rmEpiregulin, rmEGF, rmMFG-E8, recombinant human (rh) EGF, rhHB-EGF, rhHRG alpha, rhHRG beta, or recombinant rat EGF is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 463004 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG, Urogastrone | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1950 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction