missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PU.1/Spi-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7124-SP
This item is not returnable.
View return policy
Description
PU.1/Spi-1 Polyclonal specifically detects PU.1/Spi-1 in Mouse samples. It is validated for Western Blot.
Specifications
| PU.1/Spi-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hematopoietic transcription factor PU.1, OF, PU.1, SFPI1, SFPI131 kDa transforming protein, SPI1, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6688 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P17433 | |
| Spi1 | |
| E. coli-derived recombinant mouse PU.1/Spi-1 Met1-Lys169 Accession # P17433 | |
| 25 μg | |
| Primary | |
| Detects mouse PU.1/Spi-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human Spi-1 is observed, and less than 1% cross-reactivity with recombinant mouse Spi-B is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction