missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rabbit IgG VisUCyte HRP Polymer Antibody, R&D Systems™

Goat Polyclonal Antibody
€ 325.00 - € 2595.00
Specifications
| Antigen | IgG |
|---|---|
| Dilution | Immunohistochemistry |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | HRP Polymer |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18787489
![]() |
R&D Systems
VC002-025 |
25 mL | |||||||
18774708
![]() |
R&D Systems
VC002-050 |
50 mL (2 x 25 mL) |
€ 528.00
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18717428
![]() |
R&D Systems
VC002-125 |
125 mL |
€ 1360.00
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18722048
![]() |
R&D Systems
VC002-250 |
250 mL (2 x 125 mL) |
€ 2595.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgG Polyclonal specifically detects IgG in Mouse, Rabbit samples. It is validated for Immunohistochemistry.Specifications
| IgG | |
| Immunohistochemistry | |
| HRP Polymer | |
| RUO | |
| Supplied as a solution in PBS with preservative. | |
| 3500 | |
| IgG | |
| Affinity Purified | |
| Detects heavy ( gamma ) chains on mouse and rabbit IgG and light chains on all mouse and rabbit immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin mouse or rabbit serum proteins or serum proteins from bovine, horse, human, pig or rat, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC. |
| Immunohistochemistry | |
| Polyclonal | |
| Goat | |
| Mouse, Rabbit | |
| Immunoglobulin G, ImmunoglobulinG | |
| Mouse and Rabbit IgG | |
| Secondary | |
| 6 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title