missing translation for 'onlineSavingsMsg'
Learn More

Mouse Rae-1 alpha/beta/gamma Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Product Code. 18704918
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18704918 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18704918 Supplier R&D Systems Supplier No. FAB1758G100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Rae-1 Monoclonal specifically detects Rae-1 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rae-1
Applications Flow Cytometry
Classification Monoclonal
Clone 199215
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Symbols Raet1a
Host Species Rat
Immunogen BaF3 mouse pro-B cell line transfected with Rae-1 beta and Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 8480
Test Specificity Detects mouse Rae-1 alpha/beta/gamma . When tested in flow cytometry against a panel of transfectants expressing different Rae-1 proteins, this antibody recognizes mouse Rae-1 alpha , beta and gamma . It does not recognize Rae-1 delta or epsilon .
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.