missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Rae-1 alpha/beta/gamma Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1758S-100UG
This item is not returnable.
View return policy
Description
Rae-1 Monoclonal specifically detects Rae-1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Rae-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Raet1a | |
| BaF3 mouse pro-B cell line transfected with Rae-1 beta and Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta | |
| RUO | |
| 8480 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 199215 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse Rae-1 alpha/beta/gamma . When tested in flow cytometry against a panel of transfectants expressing different Rae-1 proteins, this antibody recognizes mouse Rae-1 alpha , beta and gamma . It does not recognize Rae-1 delta or epsilon . | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction