missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat Apolipoprotein H/ApoH Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6575
This item is not returnable.
View return policy
Description
Apolipoprotein H/ApoH Polyclonal specifically detects Apolipoprotein H/ApoH in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Apolipoprotein H/ApoH | |
| Polyclonal | |
| Unconjugated | |
| Q01339 | |
| APOH | |
| Mouse myeloma cell line NS0-derived recombinant mouse Apolipoprotein H/ApoH Gly20-Cys345 Accession # Q01339 | |
| 100 μg | |
| Primary | |
| Detects mouse and rat Apolipoprotein H/ApoH in Western blots and detects recombinant mouse Apolipoprotein H/ApoH in direct ELISAs. In direct ELISAs, approximately 40% cross-reactivity with recombinant human Apolipoprotein H/ApoH is observed. | |
| Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.25 ug/mL | |
| Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, APOH, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 350 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction