missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat BMP-15/GDF-9B Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF5917
This item is not returnable.
View return policy
Description
BMP-15/GDF-9B Polyclonal specifically detects BMP-15/GDF-9B in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| BMP-15/GDF-9B | |
| Polyclonal | |
| Unconjugated | |
| Q9Z0L4 | |
| BMP15 | |
| E. coli-derived recombinant mouse BMP-15/GDF-9B Gln268-Arg392 Accession # Q9Z0L4 | |
| 100 μg | |
| Primary | |
| Detects mouse and rat BMP-15/GDF-9B in Western blots and recombinant mouse in direct ELISAs. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) BMP-15 is observed and less than 1% cross-reactivity with rhBMP-3 and rhBMP-10 is observed. | |
| Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| BMP15, BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 9210 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction