missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat CD47 N-terminal IgV-like Extracellular Domain Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF1866
This item is not returnable.
View return policy
Description
CD47 Polyclonal specifically detects CD47 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CD47 | |
| Polyclonal | |
| Unconjugated | |
| NP_034711 | |
| CD47 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD47 N-terminal IgV-like Gln19-Pro158 Accession # NP_034711 | |
| 100 μg | |
| Primary | |
| Detects mouse CD47 N-terminal IgV-like in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant human CD47 is observed. | |
| Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.25-1 ug/mL, Immunohistochemistry 1-15 ug/mL | |
| antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAP, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 961 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction