missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat DARPP-32 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 7 publications
Brand: R&D Systems MAB4230
This item is not returnable.
View return policy
Description
DARPP-32 Monoclonal specifically detects DARPP-32 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| DARPP-32 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q9UD71.2 | |
| PPP1R1B | |
| E. coli-derived recombinant human DARPP-32 Ala51-Thr204 Accession # Q9UD71.2 | |
| 100 μg | |
| Primary | |
| Detects mouse and rat DARPP-32. Reactivity with human DARPP-32 has not been tested. | |
| Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 375604 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DARPP32, PPP1R1B, protein phosphatase 1, regulatory (inhibitor) subunit 1B, regulatory (inhibitor) subunit 1B (dopamine and cAMPregulated phosphoprotein, DARPP-32) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 84152 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction