missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat HO-1/HMOX1/HSP32 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF3169
This item is not returnable.
View return policy
Description
HO-1/HMOX1/HSP32 Polyclonal specifically detects HO-1/HMOX1/HSP32 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| HO-1/HMOX1/HSP32 | |
| Polyclonal | |
| Unconjugated | |
| P06762 | |
| HMOX1 | |
| E. coli-derived recombinant rat HO-1/HMOX1 Met1-Met289 Accession # P06762 | |
| 100 μg | |
| Primary | |
| Detects mouse and rat HO-1/HMOX1/HSP32 in Western blots. | |
| Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HMOX1, HO, HO1, HO-1heme oxygenase 1, HSP32 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3162 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction