missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat MMP-2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 22 publications
Brand: R&D Systems AF1488-SP
This item is not returnable.
View return policy
Description
MMP-2 Polyclonal specifically detects MMP-2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| MMP-2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72kD gelatinase, 72kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP2, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4313 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| P33434 | |
| MMP2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse MMP-2 Ile34-Cys662 Accession # P33434 | |
| 25 μg | |
| Primary | |
| Detects mouse and rat MMP-2 in direct ELISAs and Western blots. In direct ELISAs, less than 40% cross-reactivity with recombinant human MMP-2 is observed and less than 1% cross-reactivity with recombinant mouse (rm) MMP-3 and rmMMP-9 is observed. | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction