missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat MMP-2 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1488
This item is not returnable.
View return policy
Description
MMP-2 Polyclonal specifically detects MMP-2 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| MMP-2 | |
| Polyclonal | |
| Western Blot 0.1 ug/mL | |
| 72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72kD gelatinase, 72kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP2, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4313 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| Biotin | |
| P33434 | |
| MMP2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse/rat MMP-2 Ile34-Cys662 Accession # P33434 | |
| 50 μg | |
| Primary | |
| Detects mouse and rat MMP-2 in Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human MMP-2 is observed and less than 1% cross-reactivity with recombinant mouse (rm) MMP-3, rmMMP-7, and rmMMP-9 is observed. | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction