missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat MyD88 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF3109
This item is not returnable.
View return policy
Description
MyD88 Polyclonal specifically detects MyD88 in Mouse, Rat samples. It is validated for Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| MyD88 | |
| Polyclonal | |
| Unconjugated | |
| Q3U7M4 | |
| MYD88 | |
| E. coli-derived recombinant mouse MyD88 Met1-Pro296 Accession # Q3U7M4 | |
| 100 μg | |
| Primary | |
| Detects mouse and rat MyD88 in Western blots. | |
| Mouse, Rat | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4615 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction