missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat Neuropilin-1 PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems FAB566P
This item is not returnable.
View return policy
Description
Neuropilin-1 Polyclonal specifically detects Neuropilin-1 in Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Neuropilin-1 | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| BDCA4, BDCA-4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, Neuropilin1, NP1, NRP, NRP1, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8829 | |
| Mouse, Rat | |
| IgG |
| Flow Cytometry | |
| R-PE | |
| Q9QWJ9 | |
| NRP1 | |
| Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-1 Phe22-Ala810 (Lys811Arg), Ser829-Asp854 Accession # Q9QWJ9 | |
| 100 Tests | |
| Primary | |
| Detects mouse and rat Neuropilin-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 8% cross-reactivity with recombinant human Neuropilin-1 is observed, and less than 1% cross-reactivity with recombinant rat Neuropilin-2 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction