missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat PDZK1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF5780
This item is not returnable.
View return policy
Description
PDZK1 Polyclonal specifically detects PDZK1 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PDZK1 | |
| Polyclonal | |
| Unconjugated | |
| Q9JIL4 | |
| PDZK1 | |
| E. coli-derived recombinant mouse PDZK1 aa 1-519 Accession # Q9JIL4 | |
| 100 μg | |
| Primary | |
| Detects mouse and rat PDZK1 in Western blots. | |
| Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| CAP70, CFTR-associated protein of 70 kDa, CLAMP, Na(+)/H(+) exchanger regulatory factor 3, Na/Pi cotransporter C-terminal-associated protein 1, naPi-Cap1, NHERF3, NHERF-3, NHERF3PDZD1Na(+)/H(+) exchange regulatory cofactor NHE-RF3, PDZ domain containing 1, PDZ domain-containing protein 1, PDZ-containing kidney protein 1, PDZD1, Sodium-hydrogen exchanger regulatory factor 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5174 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction