missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat VSIG1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF5324
This item is not returnable.
View return policy
Description
VSIG1 Polyclonal specifically detects VSIG1 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| VSIG1 | |
| Polyclonal | |
| Biotin | |
| BAE35812 | |
| VSIG1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse VSIG1 Val23-Glu234 Accession # BAE35812 | |
| 50 μg | |
| Primary | |
| Detects mouse and rat VSIG1 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) VSIG1 is observed and less than 1% cross-reactivity with recombinant mouse (rm) VSIG2, rmVSIG3, rmVSIG4, and rhVSIG3 is observed. | |
| Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Cell surface A33 antigen, dJ889N15.1, Glycoprotein A34, GPA34, MGC44287,1700062D20Rik, V-set and immunoglobulin domain containing 1, V-set and immunoglobulin domain-containing protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 340547 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction