missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse RBP4/Retinol-Binding Protein 4 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems BAF3476
This item is not returnable.
View return policy
Description
RBP4/Retinol-Binding Protein 4 Polyclonal specifically detects RBP4/Retinol-Binding Protein 4 in Mouse samples. It is validated for Western Blot.
Specifications
| RBP4/Retinol-Binding Protein 4 | |
| Polyclonal | |
| Biotin | |
| Q00724 | |
| RBP4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse RBP4/Retinol-Binding Protein 4 Glu19-Leu201 Accession # Q00724 | |
| 50 μg | |
| Primary | |
| Detects mouse RBP4/Retinol-Binding Protein 4 in Western blots. In Western blots, approximately 15% cross-reactivity with recombinant human RBP4 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, RetinolBinding Protein 4, retinol-binding protein 4, plasma | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5950 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction