missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse RELM beta Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2730
This item is not returnable.
View return policy
Description
RELM beta Polyclonal specifically detects RELM beta in Mouse samples. It is validated for Western Blot.
Specifications
| RELM beta | |
| Polyclonal | |
| Unconjugated | |
| Q99P86 | |
| RETNLB | |
| Mouse myeloma cell line NS0-derived recombinant mouse RELM beta Gly21-Ala105 Accession # Q99P86 | |
| 100 μg | |
| Primary | |
| Detects mouse RELM beta in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant rat RELM gamma is observed and 10% cross-reactivity with recombinant mouse (rm) Resistin and rmRELM alpha is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 2, CCRG, Colon and small intestine-specific cysteine-rich protein, Colon carcinoma-related gene protein, cysteine-rich secreted A12-alpha-like protein 1, Cysteine-rich secreted protein A12-alpha-like 1, Cysteine-rich secreted protein FIZZ2, FIZZ1, FIZZ2RELM-beta, found in inflammatory zone 1, HXCP2XCP2, RELMb, RELMbeta, resistin like beta, resistin-like beta, RETNL2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 84666 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction