missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Semaphorin 3B Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5440
This item is not returnable.
View return policy
Description
Semaphorin 3B Monoclonal specifically detects Semaphorin 3B in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Semaphorin 3B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| (semaphorin) 3B, LUCA-1, SemA, Sema A(V), sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema V, SEMA3B, sema5, SEMA5FLJ34863, SEMAA, semaphorin A, semaphorin V, semaphorin-3B, Semaphorin-V, SEMAV | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7869 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| 904201 | |
| Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q62177 | |
| SEMA3B | |
| Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 3B Pro26-Pro722 Accession # Q62177 | |
| 100 μg | |
| Primary | |
| Detects mouse Semaphorin 3B in direct ELISAs. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction