missing translation for 'onlineSavingsMsg'
Learn More

Mouse Semaphorin 4D/CD100 Biotinylated Antibody, R&D Systems™

Product Code. 18130761
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18130761 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18130761 Supplier R&D Systems Supplier No. BAF5235

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Semaphorin 4D/CD100 Polyclonal specifically detects Semaphorin 4D/CD100 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Semaphorin 4D/CD100
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot
Gene Accession No. NP_038688
Gene Alias A8, BB18, CD100, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, M-sema-G, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, SEMA4D, SEMAJ, semaphorin-4D
Gene Symbols SEMA4D
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 4D/CD100 Phe24-Thr657 Accession # NP_038688
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10507
Test Specificity Detects mouse Semaphorin 4D/CD100 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) Semaphorin 3A, rhSemaphorin 4B, and rhSemaphrin 6C is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.