missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Serpin E1/PAI-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF3828
This item is not returnable.
View return policy
Description
Serpin E1/PAI-1 Polyclonal specifically detects Serpin E1/PAI-1 in Mouse samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.
Specifications
| Serpin E1/PAI-1 | |
| Polyclonal | |
| Unconjugated | |
| AAH54091 | |
| SERPINE1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Serpin E1/PAI-1 Leu24-Ser402 Accession # AAH54091 | |
| 100 μg | |
| Primary | |
| Detects mouse Serpin E1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant human Serpin E1 is observed and less than 5% cross-reactivity with recombinant mouse (rm) Serpin E2 and rmSerpin I1 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Simple Western 12.5 ug/mL, Immunoprecipitation 25 ug/mL | |
| Endothelial plasminogen activator inhibitor, Nexin, PAI1, PAI-1, PAI1PAI-1, PAISerpin E1, PLANH1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5054 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction