missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Serum Albumin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-26510-0.25MG
This item is not returnable.
View return policy
Description
Mouse Serum Albumin Polyclonal specifically detects Mouse Serum Albumin in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, SDS-Page.
Specifications
| Mouse Serum Albumin | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide | |
| Alb | |
| Anti-Mouse Serum Albumin Antibody was produced by repeated immunizations with mouse serum albumin. | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| 1 mg/mL | |
| Western Blot 1:2000 - 1:10000, ELISA 1:20000-1:100000, Immunohistochemistry 1:500 - 1:2500, SDS-Page | |
| Alb1, Alb-1, albumin 1, BCL002 | |
| Rabbit | |
| 0.25 mg | |
| 11657 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction