missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SEZ6L Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4804
This item is not returnable.
View return policy
Description
SEZ6L Polyclonal specifically detects SEZ6L in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| SEZ6L | |
| Polyclonal | |
| Unconjugated | |
| AAH65117 | |
| SEZ6L | |
| Mouse myeloma cell line NS0-derived recombinant mouse SEZ6L Glu32-Ser889 Accession # AAH65117 | |
| 100 μg | |
| Primary | |
| Detects mouse SEZ6L in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) BSRP-A and rmBSRP-C is observed. | |
| Mouse | |
| IgG |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| Acig1, AIG1, Aig1L, BSRP-B, seizure related 6 homolog (mouse)-like, seizure related gene 6-like, SEZ6L | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 23544 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction