missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SF20/MYDGF Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems IC1104V-100UG
This item is not returnable.
View return policy
Description
SF20/MYDGF Monoclonal specifically detects SF20/MYDGF in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| SF20/MYDGF | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| C19orf10, chromosome 19 open reading frame 10, EUROIMAGE1875335, IL25, IL27, IL27w, interleukin 27 working designation, interleukin-25, Ly6elg, MYDGF, myeloid-derived growth factor, R33729_1, SF20, Stromal cell-derived growth factor SF20, UPF0556 protein C19orf10 | |
| Mouse myeloma cell line NS0-derived mouse SF20/MYDGF, Val25-Leu166, Accession # Q9CPT4 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 2562A | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9CPT4 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 56005 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction