missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Siglec-1/CD169 PE-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB5610P
This item is not returnable.
View return policy
Description
Siglec-1/CD169 Monoclonal specifically detects Siglec-1/CD169 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Siglec-1/CD169 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin, Siglec1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6614 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 645608 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q62230 | |
| SIGLEC1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Siglec-1/CD169 Thr20-Leu1639 (predicted) Accession # Q62230 | |
| 100 Tests | |
| Primary | |
| Detects mouse Siglec-1/CD169 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Siglec-1/CD169, -5, -6, -7, -8, -9, -10, -11, -14, recombinant mouse Siglec-2, -3, -E, -F, -G, or -H is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction