missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Siglec-2/CD22 (NP_033975.3) VersaClone cDNA
Click to view available options
:
10 μg
Specifications
Specifications
| Gene | Siglec-2/CD22 |
| Accession Number | NP_033975.3 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2620 bp |
| Gene ID (Entrez) | 933 |
| Gene Alias | B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, Siglec2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction