missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Siglec-F Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1706-SP
This item is not returnable.
View return policy
Description
Siglec-F Monoclonal specifically detects Siglec-F in Mouse samples. It is validated for Western Blot.
Specifications
| Siglec-F | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q920G3 | |
| Siglecf | |
| Mouse myeloma cell line NS0-derived recombinant mouse Siglec-F Asp18-Thr437 (predicted) Accession # Q920G3 | |
| 25 μg | |
| Primary | |
| Detects mouse Siglec-F in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-5, -7, -9, -10, or recombinant mouse Siglec-2, or -3 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 238023 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SAF2, sialic acid binding Ig like lectin 8, SIGLEC-8, SIGLEC8L, SiglecF | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27181 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction