missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Siglec-G Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB7775U-100UG
This item is not returnable.
View return policy
Description
Siglec-G Monoclonal specifically detects Siglec-G in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Siglec-G | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_766488 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse Siglec-G in ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Siglec-F or recombinant human Siglec-10 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 805903 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Siglecg | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Siglec-G Glu20-Gln525 Accession # NP_766488 | |
| RUO | |
| 89790 | |
| Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction