missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SorCS3 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3067
This item is not returnable.
View return policy
Description
SorCS3 Polyclonal specifically detects SorCS3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.
Specifications
| SorCS3 | |
| Polyclonal | |
| Unconjugated | |
| Q8VI51 | |
| SORCS3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse SorCS3 Ala131-Ser1122 Accession # Q8VI51 | |
| 100 μg | |
| Primary | |
| Detects mouse SorCS3 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human SorCS3 is observed, approximately 7% cross-reactivity with recombinant mouse (rm) SorCS1 is observed, and less than 2% cross-reactivity with rmSorCS2 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction | |
| 6330404A12Rik, KIAA1059SORCS, sortilin-related VPS10 domain containing receptor 3, VPS10 domain receptor protein SORCS 3 (SORCS3), VPS10 domain-containing receptor SorCS3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 22986 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction