missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse STAT6 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2169
This item is not returnable.
View return policy
Description
STAT6 Monoclonal specifically detects STAT6 in Mouse samples. It is validated for Western Blot.
Specifications
| STAT6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P52633 | |
| STAT6 | |
| E. coli-derived recombinant mouse STAT6 Ser627-Trp837 Accession # P52633 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat STAT6 in Western blots. | |
| Mouse | |
| Purified |
| Western Blot | |
| 69727 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| D12S1644, EC 2.4.1.227, EC 2.7.7.6, IL-4 Stat, IL-4-STAT, signal transducer and activator of transcription 6, signal transducer and activator of transcription 6, interleukin-4 induced, STAT, interleukin-4 induced, STAT, interleukin4-induced, STAT6B, STAT6C, transcription factor IL-4 STAT | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6778 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction