missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Syndecan-3 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF2734
This item is not returnable.
View return policy
Description
SDC3 Polyclonal specifically detects SDC3 in Mouse samples. It is validated for Western Blot.
Specifications
| SDC3 | |
| Polyclonal | |
| Biotin | |
| Q64519 | |
| SDC3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Syndecan-3 Ala45-Glu384 Accession # Q64519 | |
| 50 μg | |
| Primary | |
| Detects mouse Syndecan-3 in Western blots. In Western blots, less than 2% cross-reactivity with recombinant mouse (rm) Syndecan-1, recombinant human (rh) Syndecan-2, rhSyndecan-3, and rmSyndecan-4 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| KIAA0468, N-syndecan, SDC3, SDCN, SYND3syndecan proteoglycan 3, syndecan 3, syndecan 3 (N-syndecan), syndecan neural type, syndecan-3, Syndecan3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9672 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction