missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Testican 3/SPOCK3 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF2346
This item is not returnable.
View return policy
Description
Testican 3/SPOCK3 Polyclonal specifically detects Testican 3/SPOCK3 in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| Testican 3/SPOCK3 | |
| Polyclonal | |
| Biotin | |
| Q8BKV0 | |
| SPOCK3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Testican 3/SPOCK3 Ala26-Ile436 Accession # Q8BKV0 | |
| 50 μg | |
| Primary | |
| Detects mouse Testican 3/SPOCK3 in ELISAs and Western blots. In sandwich immunoassays, less than 0.3% cross-reactivity with recombinant human (rh) Testiscan 1 and rhTestican2 is observed. | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| CWCV, and Kazal-like domains proteoglycan 3, HSAJ1454, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3, SPOCK3, TES-3, testican 3, TICN3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 50859 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction