missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TLR2 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1530V-100UG
This item is not returnable.
View return policy
Description
TLR2 Monoclonal specifically detects TLR2 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| TLR2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9QUN7 | |
| TLR2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TLR2 Gln25-Leu590 Accession # Q9QUN7 | |
| RUO | |
| 7097 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 203325 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD282, CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse TLR2 in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant mouse (rm) TLR1, recombinant human (rh) TLR2, rhTLR3, rhTLR4, or rmTLR6. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction