missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TNF-alpha PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC410P
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| TNF-alpha | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| TNF | |
| Mouse TNF-alpha | |
| 100 Tests | |
| Primary | |
| Detects mouse TNF-alpha in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| MP6-XT22 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7124 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction