missing translation for 'onlineSavingsMsg'
Learn More

Mouse TRAIL/TNFSF10 Biotinylated Antibody, R&D Systems™

Product Code. 18013202
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18013202 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18013202 Supplier R&D Systems Supplier No. BAF1121

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

TRAIL/TNFSF10 Polyclonal specifically detects TRAIL/TNFSF10 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Detection (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAIL/TNFSF10
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. P50592
Gene Alias Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TNFSF10, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10
Gene Symbols TNFSF10
Host Species Goat
Immunogen E. coli-derived recombinant mouse TRAIL/TNFSF10 Pro118-Asn291 Accession # P50592
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8743
Test Specificity Detects mouse TRAIL/TNFSF10 in ELISAs and Western blots. In sandwich ELISAs, less than 0.1% cross-reactivity with recombinant human TRAIL is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.