missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VE-Cadherin Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB1002
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
VE-Cadherin Monoclonal specifically detects VE-Cadherin in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Spezifikation
| VE-Cadherin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| 2208309A | |
| CDH5 | |
| Mouse myeloma cell line NS0-derived recombinant mouse VE-Cadherin Asp46-Gln592 (Gly67 del, Ile69Asp, Lys70Gln) Accession # 2208309A | |
| 100 μg | |
| Primary | |
| Detects mouse VE-Cadherin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cadherin-8, rhCadherin-17, rhN-Cadherin, recombinant mouse (rm) E-Cadherin, or rmP-Cadherin is observed. | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 162709 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, CDH5, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin, VECadherin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1003 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur