missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VEGF120/164 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems MAB493
This item is not returnable.
View return policy
Description
VEGF Monoclonal specifically detects VEGF in Mouse samples. It is validated for Western Blot.
Specifications
| VEGF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7422 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 39917 | |
| Western Blot 1 ug/mL | |
| AAA40547 | |
| VEGFA | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse VEGF 164 Ala27-Arg190 Accession # AAA40547 | |
| 500 μg | |
| Primary | |
| Detects mouse VEGF 120/164 in direct ELISAs and Western blots. This antibody does not recognize recombinant mouse VEGF 115 or any isoform of human VEGF. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction