missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VISTA/B7-H5/PD-1H Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB70051
This item is not returnable.
View return policy
Description
VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| VISTA/B7-H5/PD-1H | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q9D659 | |
| Vsir | |
| Mouse myeloma cell line NS0-derived recombinant mouse VISTA/B7-H5/PD-1H Phe33-Ala191 Accession # Q9D659 | |
| 100 μg | |
| Primary | |
| Detects mouse VISTA/B7-H5/PD-1H in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human VISTA/B7-H5/PD-1H is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 742002 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 4632428N05Rik, B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, Dies1, Gi24, PD1H, PD-1H, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64115 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction