Learn More
Mouse Vitamin D BP Antibody, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody
Brand:  R&D Systems AF4188-SP
Product Code. 18746427
  • € 188.00 / 25µg
Description
Vitamin D BP Polyclonal specifically detects Vitamin D BP in Mouse samples. It is validated for Western Blot, Simple Western.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vitamin D BP
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
DBP, DBP/GC, DBP-MAF, DBPVDBG, EC 6.3.1.5, Gc, Gc-globulin, Gc-MAF, GRD3, Group-specific component, group-specific component (vitamin D binding protein), hDBP, VDB, VDBG, VDBP, vitamin D-binding alpha-globulin, vitamin D-binding protein
Sheep
Affinity Purified
RUO
2638
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL
P21614
GC
Mouse myeloma cell line NS0-derived recombinant mouse Vitamin D BP Leu17-Ser476 Accession # P21614
25 μg
Primary
Detects mouse Vitamin D BP in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human Vitamin D BP is observed.