missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Vitronectin Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 7 publications
Brand: R&D Systems MAB38751
This item is not returnable.
View return policy
Description
Vitronectin Monoclonal specifically detects Vitronectin in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| Vitronectin | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| AAH12690 | |
| VTN | |
| Mouse myeloma cell line NS0-derived recombinant mouse Vitronectin Asp20-Lys478 Accession # AAH12690 | |
| 100 μg | |
| Primary | |
| Detects mouse Vitronectin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Vitronectin is observed. | |
| Mouse | |
| Purified |
| Immunohistochemistry | |
| 347317 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT, VTN | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7448 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction