Learn More
R&D Systems™ Mouse XL Cytokine Luminex™ Performance Premixed Kit
Product Code. 18744707
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
23 Plex
24 Plex
25 Plex
26 Plex
27 Plex
28 Plex
29 Plex
3 Plex
30 Plex
31 Plex
32 Plex
33 Plex
34 Plex
35 Plex
36 Plex
37 Plex
38 Plex
39 Plex
4 Plex
40 Plex
41 Plex
42 Plex
43 Plex
44 Plex
45 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
Each
Product Code. 18744707
Brand: R&D Systems™ FCSTM2018
Specifications
Quantity 18 Plex
Species Mouse
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.