Learn More
R&D Systems™ Mouse XL VEGF, Luminex™ Performance Assay
Product Code. 18441910
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 18441910
Brand: R&D Systems™ LMXL493
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Species Mouse
Gene ID (Entrez) 7422
Synonym MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.