missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOX1 Antibody (OTI5B11), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72770F
This item is not returnable.
View return policy
Description
MOX1 Monoclonal specifically detects MOX1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| MOX1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| MEOX1 | |
| Full length recombinant protein of human MOX1 (NP_004518) produced in E.coli. | |
| 0.1 mL | |
| 4222.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI5B11 | |
| Immunohistochemistry | |
| homeobox protein MOX-1, mesenchyme homeobox 1MOX1mesenchyme homeo box 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction