Learn More
MP Biomedicals™ 5-Fluoroorotic Acid [ 5-FOA] (POWDER) Monohydrate
Pale yellow crystalline powder with applications in yeast cell culture
Brand:  MP Biomedicals™ 114066102
Additional Details : Weight : 0.01000kg
Product Code. 11367849
  • € 489.72 / 1g
Estimated Shipment: 16-04-2024
to see stock.
Description
Description
  • Physical Description: Pale Yellow Crystalline Powder
  • Formula: C5H3FN2O4H2O
  • CAS #: 703-95-7
  • Formula Weight: 174.09 (anhy)
Yeast Cell Culture
Specifications
Specifications
1 g