Learn More
MP Biomedicals™ BCIP/NBT Liq. Substrate Plus, 500mL
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Chromagen Type | BCIP or NBT |
| Form | Liquid |
| For Use With (Application) | For visualizing alkaline phosphatase activity in immunoblotting and immunohistology |
| Quantity | 500 mL |
| Substrate Type | BCIP/NBT Substrate |
| pH | 9.8 |
| Storage Requirements | 2° to 8°C |
| Substrate Quantity | 500mL |