Learn More
MP Biomedicals™ Bilirubin Oxidase
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 U
100 U
Unit Size:
10 units
100 units
Description
Activity: ≥0.8U/mg solid. Unit Definition: One unit is the amount of enzyme that oxidizes one micromole of bilirubin per minute at 37°C, pH 8.4. Lyophilized powder.
Specifications
Specifications
| Concentration | ≥0.8 U/mg |
| Color | Light Blue/Green |
| For Use With (Application) | Degrade Bilirubin, used to determine free hemoglobin in icteric specimens, also has potential application in dye effluent decolorization and is a potential treatment for neonatal jaundice |
| Content And Storage | -20°C |
| Source | Myrothecium species |
| Quantity | 100 U |
| Product Type | Bilirubin Oxidase |
| Molecular Weight (g/mol) | 52000 |
| Form | Lyophilized Powder |