Learn More
MP Biomedicals™ Bilirubin Oxidase
Product Code. p-3563345 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 U
100 U
Unit Size:
10 units
100 units
Product Code. 11404462
Brand: MP Biomedicals™ 0219000080
From Myrothecium species
Activity: ≥0.8U/mg solid. Unit Definition: One unit is the amount of enzyme that oxidizes one micromole of bilirubin per minute at 37°C, pH 8.4. Lyophilized powder.
TRUSTED_SUSTAINABILITY
Specifications
Concentration ≥0.8 U/mg
Color Light Blue/Green
For Use With (Application) Degrade Bilirubin, used to determine free hemoglobin in icteric specimens, also has potential application in dye effluent decolorization and is a potential treatment for neonatal jaundice
Content And Storage -20°C
Source Myrothecium species
Quantity 100 U
Product Type Bilirubin Oxidase
Molecular Weight (g/mol) 52000
Form Lyophilized Powder